3-Methylbenzyl isothiocyanate ≥99%
Supplier: Matrix Scientific
056596-1G
056596-5G
101910-990EA
84.62
USD
101910-990
101910-992
3-Methylbenzyl isothiocyanate ≥99%
3-Methylbenzyl isothiocyanate
Hazard: Irritant-Harmful
Formula:
C₉H₉NS Boiling Pt: 127°C/2mm Density: 1.09 |
MDL Number:
MFCD00060410 CAS Number: 3696-66-0 |
Specification Test Results
Boiling point | 127°C/2mm |
Density | 1.09 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...