2-Furoyl isothiocyanate

Supplier: Matrix Scientific

056565-1G 056565-5G
101910-892EA 87.69 USD
101910-892 101910-894
2-Furoyl isothiocyanate
2-Furoyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₆H₃NO₂S
MW: 153.16 g/mol
Boiling Pt: 78 °C (1 mmHg)
Density: 1.3
MDL Number: MFCD00060332
CAS Number: 80440-95-5

Order Now

Specification Test Results

Boiling point 78°C/1mm
Density 1.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR