Ethyl L-2-isothiocyanatopropionate

Supplier: Matrix Scientific

056563-1G 056563-5G
101910-886EA 70.77 USD
101910-886 101910-888
Ethyl L-2-isothiocyanatopropionate
Ethyl L-2-isothiocyanatopropionate

Hazard: Irritant-Harmful

Formula: C₆H₉NO₂S
Boiling Pt: 44 °C (1.5 mmHg)
Density: 1.09
MDL Number: MFCD09025691
CAS Number: 39574-16-8

Order Now

Specification Test Results

Boiling point 44°C/1.5mm
Density 1.09


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR