Ethyl DL-3-isothiocyanatobutyrate ≥99%

Supplier: Matrix Scientific

056561-1G 056561-5G
101910-880EA 103.08 USD
101910-880 101910-882
Ethyl DL-3-isothiocyanatobutyrate ≥99%
Ethyl DL-3-isothiocyanatobutyrate

Hazard: Irritant-Harmful

Formula: C₇H₁₁NO₂S
Boiling Pt: 88°C/5mm
Density: 1.07
MDL Number: MFCD00041107
CAS Number: 206750-29-0

Order Now

Specification Test Results

Boiling point 88°C/5mm
Density 1.07

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR