Ethyl-3-isothiocyanatobenzoate ≥99%

Supplier: Matrix Scientific

056559-5G 056559-1G
101910-876EA 267.69 USD
101910-876 102551-428
Ethyl-3-isothiocyanatobenzoate ≥99%
Ethyl 3-isothiocyanatobenzoate

Hazard: Irritant-Harmful

Formula: C₁₀H₉NO₂S
Boiling Pt: 112°C/1mm
Density: 1.18
MDL Number: MFCD00060375
CAS Number: 3137-84-6

Order Now

Specification Test Results

Boiling point 112°C/1mm
Density 1.18

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR