Ethyl-8-bromooctanoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 8-Bromooctanoate

007592-5G 007592-25G
101793-072EA 21.54 USD
101793-072 101793-074
Ethyl-8-bromooctanoate ≥97%
Ethyl-8-bromooctanoate

Hazard: Irritant

Formula: C₁₀H₁₉BrO₂
MW: 251.17 g/mol
Boiling Pt: 123 °C ( 3 mmHg)
Density: 1.194
MDL Number: MFCD00045057
CAS Number: 29823-21-0

Order Now

Specification Test Results

Boiling point 123°C/ 3mm
Density 1.194

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR