4-Methoxy-2-methylbenzonitrile ≥97%

Supplier: Matrix Scientific

007474-5G 007474-25G
101792-876EA 73.85 USD
101792-876 101792-878
4-Methoxy-2-methylbenzonitrile ≥97%
4-Methoxy-2-methylbenzonitrile

Hazard: Irritant/Store in Freezer

Formula: C₉H₉NO
MW: 147.1700 g/mol
Boiling Pt: 104°C/5mm
Density: 1.06
MDL Number: MFCD03425692
CAS Number: 21883-13-6

Order Now

Specification Test Results

Boiling point 104°C/5mm
Density 1.06

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR