1,3-Dichloro-1,1-difluoropropane ≥98%
Supplier: Matrix Scientific
007194-5G
007194-25G
007194-100G
101792-156EA
63.08
USD
101792-156
101792-158
101792-160
1,3-Dichloro-1,1-difluoropropane ≥98%
1,3-Dichloro-1,1-difluoropropane
TCA listed: F
Hazard: Ozone Depleter
Formula:
C₃H₄Cl₂F₂ Boiling Pt: 81…81.5 °C Density: 1.339 |
MDL Number:
MFCD01320726 CAS Number: 819-00-1 |
Specification Test Results
Boiling point | 81-81.5°C |
Density | 1.339 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...