1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane ≥97%
Supplier: Matrix Scientific
007115-5G
007115-25G
101792-052EA
150.77
USD
101792-052
101792-054
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane ≥97%
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane
Hazard: Flammable/Corrosive/Moisture Sensitive
Formula:
C₉H₇Cl₂F₁₃Si MW: 461.13 g/mol Boiling Pt: 76 °C (12 mmHg) Density: 1.55 |
MDL Number:
MFCD00042360 CAS Number: 73609-36-6 UN: 2920 ADR: 8,II |
Specification Test Results
Boiling point | 76°C/12mm |
Density | 1.55 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...