Bis(perfluoroisopropyl)ketone ≥97%

Supplier: Matrix Scientific

Synonyms: Bis(perfluoroisopropyl)methanone

006480-1G 006480-5G
101790-592EA 73.85 USD
101790-592 101790-594
Bis(perfluoroisopropyl)ketone ≥97%
Bis(perfluoroisopropyl)ketone

Hazard: Irritant

Formula: C₇F₁₄O
MW: 366.06 g/mol
Boiling Pt: 72-73°C
Density: 1.661
MDL Number: MFCD00042087
CAS Number: 813-44-5

Order Now

Specification Test Results

Boiling point 72-73°C
Density 1.661

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR