4-(Methylthio)benzylamine
Supplier: Matrix Scientific
Synonyms:
(4-(Methylthio)phenyl)methanamine
006304-1G
006304-5G
101790-366EA
135.38
USD
101790-366
101790-368
4-(Methylthio)benzylamine
4-(Methylthio)benzylamine
Hazard: Irritant/Stench
Formula:
C₈H₁₁NS Boiling Pt: 114°C/5mm Density: 1.1 |
MDL Number:
MFCD01310831 CAS Number: 83171-39-5 |
Specification Test Results
Boiling point | 114°C/5mm |
Density | 1.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...