1-Chloro-4-propylbenzene

Supplier: Matrix Scientific

Synonyms: 4-Propylchlorobenzene

006150-1G 006150-5G
101790-156EA 56.92 USD
101790-156 101790-158
1-Chloro-4-propylbenzene
1-Chloro-4-propylbenzene

Hazard: Irritant

Formula: C₉H₁₁Cl
MW: 154.64 g/mol
Boiling Pt: 111 °C (5 mmHg)
Density: 1.01
MDL Number: MFCD00270099
CAS Number: 52944-34-0

Order Now

Specification Test Results

Boiling point 111°C/5mm
Density 1.01

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR