3-(2-Chlorophenyl)propiononitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(2-Chlorophenyl)propanenitrile

006113-1G 006113-5G
101790-122EA 38.46 USD
101790-122 101790-124
3-(2-Chlorophenyl)propiononitrile ≥97%
3-(2-Chlorophenyl)propiononitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₈ClN
MW: 165.6200 g/mol
Boiling Pt: 113 °C (5 mmHg)
Density: 1.14
MDL Number: MFCD00019873
CAS Number: 7315-17-5

Order Now

Specification Test Results

Boiling point 113°C/5mm
Density 1.14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR