2-Bromodiphenylmethane ≥98%

Supplier: Matrix Scientific

005721-1G 005721-5G
101789-440EA 96.92 USD
101789-440 101789-442
2-Bromodiphenylmethane ≥98%
2-Bromodiphenylmethane

Hazard: Irritant

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Boiling Pt: 109 °C (1 mmHg)
Density: 1.34
MDL Number: MFCD00079713
CAS Number: 23450-18-2

Order Now

Specification Test Results

Boiling point 109°C/1mm
Density 1.34

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR