4-Bromodiphenylmethane

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-4-bromobenzene

005720-1G 005720-5G
101789-436EA 52.31 USD
101789-436 101789-438
4-Bromodiphenylmethane
4-Bromodiphenylmethane

Hazard: Irritant

Formula: C₁₃H₁₁Br
MW: 247.130 g/mol
Boiling Pt: 124 °C (4 mmHg)
Density: 1.34
MDL Number: MFCD00053165
CAS Number: 2116-36-1

Order Now

Specification Test Results

Boiling point 124°C/4mm
Density 1.34

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR