3-(4-Methoxyphenyl)propiononitrile ≥99%

Supplier: Matrix Scientific

005666-1G 005666-5G
101789-310EA 63.08 USD
101789-310 101789-312
3-(4-Methoxyphenyl)propiononitrile ≥99%
3-(4-Methoxyphenyl)propiononitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₀H₁₁NO
Boiling Pt: 105°C/1mm
Density: 1.05
MDL Number: MFCD00016404
CAS Number: 22442-48-4

Order Now

Specification Test Results

Boiling point 105°C/1mm
Density 1.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR