2-(2-Hydroxyethyl)benzotrifluoride ≥97%

Supplier: Matrix Scientific

005470-5G 005470-25G
101789-008EA 121.54 USD
101789-008 101789-010
2-(2-Hydroxyethyl)benzotrifluoride ≥97%
2-(2-Hydroxyethyl)benzotrifluoride

Hazard: Irritant

Formula: C₉H₉F₃O
Boiling Pt: 184…186 °C
Density: 1.197
MDL Number: MFCD00003540
CAS Number: 94022-96-5

Order Now

Specification Test Results

Boiling point 184-186°C
Density 1.197


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR