5-Chloro-1-methylimidazole

Supplier: Matrix Scientific

032734-1G 032734-5G
101834-328EA 73.85 USD
101834-328 101834-330
5-Chloro-1-methylimidazole
5-Chloro-1-methylimidazole

Hazard: Irritant

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
MDL Number: MFCD00014505
CAS Number: 872-49-1

Order Now

Specification Test Results

Boiling point 82-85 ºC
Density 1.25

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR