3-(Cyclopropylamino)propionitrile ≥99%

Supplier: Matrix Scientific

004959-1G 004959-5G
101788-084EA 143.08 USD
101788-084 101788-086
3-(Cyclopropylamino)propionitrile ≥99%
3-(Cyclopropylamino)propionitrile

Hazard: Irritant

Formula: C₆H₁₀N₂
Boiling Pt: 115 °C (20 mmHg)
Density: 0.96
MDL Number: MFCD00060285
CAS Number: 58196-47-7

Order Now

Specification Test Results

Boiling point 115°C/20mm
Density 0.96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR