3-(3-Bromophenyl)propiononitrile ≥98%
Supplier: Matrix Scientific
004942-1G
004942-5G
101788-072EA
69.23
USD
101788-072
101788-074
3-(3-Bromophenyl)propiononitrile ≥98%
3-(3-Bromophenyl)propiononitrile
Hazard: Irritant/Irritant-Harmful
Formula:
C₉H₈BrN Boiling Pt: 113 °C (1 mmHg) Density: 1.42 |
MDL Number:
MFCD01310794 CAS Number: 376646-63-8 |
Specification Test Results
Boiling point | 113°C/1mm |
Density | 1.42 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...