Methyl (3-methoxyphenyl)acetate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Methoxyphenylacetate

004467-5G 004467-25G
101787-284EA 70.77 USD
101787-284 101787-286
Methyl (3-methoxyphenyl)acetate ≥98%
Methyl (3-methoxyphenyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.23 g/mol
Boiling Pt: 130°C/20mm
Density: 1.12
MDL Number: MFCD00017205
CAS Number: 18927-05-4

Order Now

Specification Test Results

Boiling point 130°C/20mm
Density 1.12

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR