3-(Dodecylamino)propionitrile
Supplier: Matrix Scientific
004274-5G
004274-25G
101787-038EA
44.62
USD
101787-038
101787-040
3-(Dodecylamino)propionitrile
3-(Dodecylamino)propionitrile
Hazard: Irritant/Irritant-Harmful
Formula:
C₁₅H₃₀N₂ Boiling Pt: 140°C/1mm Density: 0.86 |
MDL Number:
MFCD00060312 CAS Number: 4763-40-0 |
Specification Test Results
Boiling point | 140°C/1mm |
Density | 0.86 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...