3,4-Dimethylphenethylamine ≥98%

Supplier: Matrix Scientific

004083-1G 004083-5G
101786-806EA 89.23 USD
101786-806 101786-808
3,4-Dimethylphenethylamine ≥98%
3,4-Dimethylphenethylamine

Hazard: Irritant

Formula: C₁₀H₁₅N
Boiling Pt: 110 °C (15 mmHg)
Density: 0.94
CAS Number: 17283-14-6

Order Now

Specification Test Results

Boiling point 110°C/15mm
Density 0.94


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR