2,4-Dimethylbenzylamine ≥98%
Supplier: Matrix Scientific
004074-5G
004074-25G
101786-788EA
320
USD
101786-788
101786-790
2,4-Dimethylbenzylamine ≥98%
2,4-Dimethylbenzylamine
Hazard: Irritant/Air Sensitive
Formula:
C₉H₁₃N Boiling Pt: 108 °C (20 mmHg) Density: 0.96 |
MDL Number:
MFCD00025575 CAS Number: 94-98-4 |
Specification Test Results
Boiling point | 108°C/20mm |
Density | 0.96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...