4-Methylphenylacetone ≥98%
Supplier: Matrix Scientific
Synonyms:
1-(p-Tolyl)propan-2-one
003903-5G
003903-25G
101786-402EA
69.23
USD
101786-402
101786-404
4-Methylphenylacetone ≥98%
4-Methylphenylacetone
Hazard: Irritant/Lachrymator/Store in Freezer
Formula:
C₁₀H₁₂O MW: 148.20 g/mol Boiling Pt: 132 °C (20 mmHg) Density: 0.96 |
MDL Number:
MFCD00017250 CAS Number: 2096-86-8 |
Specification Test Results
Boiling point | 132°C/20mm |
Density | 0.96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...