3-(Cyclohexylamino)propionitrile ≥98%

Supplier: Matrix Scientific

003531-5G 003531-25G
101785-846EA 33.85 USD
101785-846 101785-848
3-(Cyclohexylamino)propionitrile ≥98%
3-(Cyclohexylamino)propionitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₁₆N₂
Boiling Pt: 122…124 °C (4 mmHg)
Density: 0.94
MDL Number: MFCD00021325
CAS Number: 702-03-4

Order Now

Specification Test Results

Boiling point 122-124°C/4
Density 0.94

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR