3-(Cyclooctylamino)propionitrile ≥98%

Supplier: Matrix Scientific

003118-1G 003118-5G
101785-270EA 30.77 USD
101785-270 101785-272
3-(Cyclooctylamino)propionitrile ≥98%
3-(Cyclooctylamino)propionitrile

Hazard: Irritant

Formula: C₁₁H₂₀N₂
Boiling Pt: 124°C/1mm
Density: 0.95
MDL Number: MFCD00060288
CAS Number: 206559-53-7

Order Now

Specification Test Results

Boiling point 124°C/1mm
Density 0.95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR