4-Phenoxybenzylamine ≥98%

Supplier: Matrix Scientific

003029-5G 003029-1G
101785-106EA 144.62 USD
101785-106 102832-754
4-Phenoxybenzylamine ≥98%
4-Phenoxybenzylamine

Hazard: Irritant/Irritant-Harmful

Formula: C₁₃H₁₃NO
Boiling Pt: 124 °C (1 mmHg)
Density: 1.12
MDL Number: MFCD01310836
CAS Number: 107622-80-0

Order Now

Specification Test Results

Boiling point 124°C/1mm
Density 1.12


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR