4-Butylbenzonitrile ≥98%

Supplier: Matrix Scientific

003021-5G 003021-25G
101785-098EA 103.08 USD
101785-098 101785-100
4-Butylbenzonitrile ≥98%
4-Butylbenzonitrile

Hazard: Irritant-Harmful

Formula: C₁₁H₁₃N
Boiling Pt: 102°C/2mm
Density: 0.94
MDL Number: MFCD00052345
CAS Number: 20651-73-4

Order Now

Specification Test Results

Boiling point 102°C/2mm
Density 0.94


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR