2,2,2-Trifluoroethyl crotonate ≥99% inhibited

Supplier: Matrix Scientific


Stabilization: <250 ppm Stabilised with 4-methoxyphenol (MEHQ)

009188-1G 009188-5G
101796-600EA 44.62 USD
101796-600 101796-602
2,2,2-Trifluoroethyl crotonate ≥99% inhibited
2,2,2-Trifluoroethyl crotonate

Inhibited with MEHQ, <250 ppm


Hazard: Flammable

Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 123…124 °C
Density: 1.18
Flash Pt: <37 °C
MDL Number: MFCD04038364
CAS Number: 287957-36-2

Order Now

Specification Test Results

Boiling point 52-53°C/115
Density 1.18


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR