1,1,2-Trichloro-1,3,3,3-tetrafluoropropane ≥97%

Supplier: Matrix Scientific

009180-1G 009180-5G
101796-580EA 93.85 USD
101796-580 101796-582
1,1,2-Trichloro-1,3,3,3-tetrafluoropropane ≥97%
1,1,2-Trichloro-1,3,3,3-tetrafluoropropane

Hazard: Ozone Depleter

Formula: C₃HCl₃F₄
Boiling Pt: 87…87.3 °C
Density: 1.6174
MDL Number: MFCD04038362
CAS Number: 431-84-5

Order Now

Specification Test Results

Boiling point 87-87.3°C
Density 1.617


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR