Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride ≥90%

Supplier: Matrix Scientific

009103-1G 009103-5G
101796-350EA 78.46 USD
101796-350 101796-352
Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride ≥90%
Perfluoro-2,5,8,11,14,17,20-heptamethyl-3,6,9,12,15,18,21-heptaoxatetracosanoyl fluoride

Balance is other HFPO oligomer acid fluorides


Hazard: Corrosive/Irritant

Formula: C₂₄F₄₈O₈
Boiling Pt: 120…122 °C (1 mmHg)
Density: 1.8
MDL Number: MFCD03411205
CAS Number: 13140-25-5

Order Now

Specification Test Results

Boiling point 120-122°C/1
Density 1.8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR