Hexafluoroisopropyl crotonate ≥99%

Supplier: Matrix Scientific

008991-1G 008991-5G
101796-030EA 78.46 USD
101796-030 101796-032
Hexafluoroisopropyl crotonate ≥99%
Hexafluoroisopropyl crotonate

Hazard: Flammable/Irritant

Formula: C₇H₆F₆O₂
Boiling Pt: 62-63°C/115mm
Density: 1.3
MDL Number: MFCD02093322
CAS Number: 135771-94-7

Order Now

Specification Test Results

Boiling point 62-63°C/115
Density 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR