1,1-Dichloro-2,2-difluoroethane ≥97%

Supplier: Matrix Scientific

008893-1G 008893-5G
101795-736EA 73.85 USD
101795-736 101795-738
1,1-Dichloro-2,2-difluoroethane ≥97%
1,1-Dichloro-2,2-difluoroethane

Hazard: Ozone Depleter/Irritant/Refrigerate

Formula: C₂H₂Cl₂F₂
Boiling Pt: 60 °C
Density: 1.4945
MDL Number: MFCD02094735
CAS Number: 471-43-2

Order Now

Specification Test Results

Boiling point 60°C
Density 1.4945

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR