3-Ethoxy-4-methoxyphenethylamine ≥99%

Supplier: Matrix Scientific

002562-1G 002562-5G
101784-168EA 73.85 USD
101784-168 101784-170
3-Ethoxy-4-methoxyphenethylamine ≥99%
3-Ethoxy-4-methoxyphenethylamine

Hazard: Irritant/Irritant-Harmful

Formula: C₁₁H₁₇NO₂
Boiling Pt: 119 °C (1 mmHg)
Density: 1.05
MDL Number: MFCD00079755
CAS Number: 86456-97-5

Order Now

Specification Test Results

Boiling point 119°C/1mm
Density 1.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR