2-Chloro-4'-fluoropropiophenone ≥98%

Supplier: Matrix Scientific

002408-1G 002408-5G
101783-856EA 36.92 USD
101783-856 101783-858
2-Chloro-4'-fluoropropiophenone ≥98%
2-Chloro-4'-fluoropropiophenone

Hazard: Irritant

Formula: C₉H₈ClFO
Boiling Pt: 71°C/1mm
Density: 1.24
MDL Number: MFCD00060275
CAS Number: 81112-09-6

Order Now

Specification Test Results

Boiling point 71°C/1mm
Density 1.24

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR