3-Bromo-4-methoxyphenethylamine ≥98%

Supplier: Matrix Scientific

001800-1G 001800-5G
101782-620EA 136.92 USD
101782-620 101782-622
3-Bromo-4-methoxyphenethylamine ≥98%
3-Bromo-4-methoxyphenethylamine

Hazard: Irritant

Formula: C₉H₁₂BrNO
MW: 230.11 g/mol
Boiling Pt: 114 °C (1 mmHg)
Density: 1.41
MDL Number: MFCD00153891
CAS Number: 159465-27-7

Order Now

Specification Test Results

Boiling point 114°C/1mm
Density 1.41

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR