(3-Methoxy-4-methylphenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Methoxy-4-methylphenylacetonitrile

001495-1G 001495-5G
101781-814EA 90.77 USD
101781-814 101781-816
(3-Methoxy-4-methylphenyl)acetonitrile ≥98%
(3-Methoxy-4-methylphenyl)acetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₀H₁₁NO
MW: 161.2004 g/mol
Boiling Pt: 176 °C (25 mmHg)
Density: 1.07
MDL Number: MFCD00060309
CAS Number: 64829-31-8

Order Now

Specification Test Results

Boiling point 176°C/25mm
Density 1.07


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR