4-Methoxy-3-methylphenylacetonitrile ≥98%

Supplier: Matrix Scientific

001463-1G 001463-5G
101781-724EA 87.69 USD
101781-724 101781-726
4-Methoxy-3-methylphenylacetonitrile ≥98%
4-Methoxy-3-methylphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₀H₁₁NO
Boiling Pt: 127°C/2mm
Density: 1.06
MDL Number: MFCD00040891
CAS Number: 75391-57-0

Order Now

Specification Test Results

Boiling point 127°C/2mm
Density 1.06

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR