Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride ≥97%

Supplier: Matrix Scientific

001417-5G 001417-25G 001417-100G
101781-612EA 58.46 USD
101781-612 101781-614 101781-616
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride ≥97%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoyl fluoride

TCA listed: T
Hazard: Corrosive

Formula: C₁₂F₂₄O₄
Boiling Pt: 114…116 °C
Density: 1.8
MDL Number: MFCD00084743
CAS Number: 27639-98-1

Order Now

Specification Test Results

Boiling point 158-161°C
Density 1.8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR