4'-Decyl-2,2,2-trifluoroacetophenone

Supplier: Matrix Scientific

001410-1G 001410-5G
101781-598EA 153.85 USD
101781-598 101781-600
4'-Decyl-2,2,2-trifluoroacetophenone
4'-Decyl-2,2,2-trifluoroacetophenone

Hazard: Irritant-Harmful

Formula: C₁₈H₂₅F₃O
Boiling Pt: 120 °C (1 mmHg)
Density: 1.04
MDL Number: MFCD00060688
CAS Number: 100444-41-5

Order Now

Specification Test Results

Boiling point 120°C/1mm
Density 1.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR