Perfluoro-15-crown-5-ether 99

Supplier: Matrix Scientific

Synonyms: Icosafluoro-15-crown 5-Ether, Icosafluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-15-crown 5-Ether

001399-1G 001399-5G
101781-560EA 230.77 USD
101781-560 101781-562
Perfluoro-15-crown-5-ether 99
Perfluoro-15-crown-5-ether

Hazard: Irritant

Formula: C₁₀F₂₀O₅
MW: 580.08 g/mol
Boiling Pt: 145 °C
Density: 1.3
MDL Number: MFCD01320747
CAS Number: 97571-69-2

Order Now

Specification Test Results

Boiling point 145°C
Density >1.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR