1-Iodo-1-(trifluoromethyl)tetrafluoroethane ≥97%

Supplier: Matrix Scientific

001110-5G 001110-25G 001110-100G
101780-982EA 95.38 USD
101780-982 101780-984 101780-986
1-Iodo-1-(trifluoromethyl)tetrafluoroethane ≥97%
1-Iodo-1-(trifluoromethyl)tetrafluoroethane

Hazard: Irritant

Formula: C₃F₇IO
MW: 311.92 g/mol
Boiling Pt: 43…44 °C
Density: 1.6
MDL Number: MFCD00077594
CAS Number: 69066-98-4

Order Now

Specification Test Results

Boiling point 43-44°C
Density >1.6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR