Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate ≥97%

Supplier: Matrix Scientific

001091-25G 001091-100G
101780-928EA 44.62 USD
101780-928 101780-930
Methyl-2,2-difluoro-2-(fluorosulfonyl)acetate ≥97%
Methyl 2,2-difluoro-2-(fluorosulphonyl)acetate

Hazard: Flammable/Corrosive/Moisture Sensitive

Formula: C₃H₃F₃O₄S
MW: 192.11
Boiling Pt: 118°C
Density: 1.52
MDL Number: MFCD00144316
CAS Number: 680-15-9

Order Now

Specification Test Results

Boiling point 118°C
Density 1.52

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR