2,5-Dichloropyridine-6-carboxaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3,6-Dichloropicolinaldehyde

056292-1G 056292-5G
101909-954EA 61.54 USD
101909-954 101909-956
2,5-Dichloropyridine-6-carboxaldehyde ≥97%
2,5-Dichloropyridine-6-carboxaldehyde

Hazard: Irritant

Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Boiling Pt: 234°C
Density: 1.489
Flash Pt: 95°C
MDL Number: MFCD12963768
CAS Number: 343781-53-3

Order Now

Specification Test Results

Boiling point 234°C
Density 1.489
ND20 1.608

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR