5,5-Dimethyl-1-pyrroline-N-oxide ≥98%

Supplier: Matrix Scientific

042487-1G 042487-5G 042487-25G
101855-996EA 63.08 USD
101855-996 101855-998 101856-000
5,5-Dimethyl-1-pyrroline-N-oxide ≥98%
5,5-Dimethyl-1-pyrroline-N-oxide

Hazard: Irritant/Air Sensitive/Hygroscopic/Store in Freezer

Formula: C₆H₁₁NO
Boiling Pt: 75 °C (0.4 mmHg)
Melting Pt: 25…29 °C
Density: 1.015
MDL Number: MFCD00005279
CAS Number: 3317-61-1

Order Now

Specification Test Results

Boiling point 75°C/0.4mm
Density 1.015
ND20 1.496


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR