Ethyl phenylglyoxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl benzoylformate
024775-5G
024775-25G
101818-874EA
24.62
USD
101818-874
101818-876
Ethyl phenylglyoxylate ≥97%
Ethyl phenylglyoxylate
Hazard: Irritant
Formula:
C₁₀H₁₀O₃ MW: 178.19 g/mol Boiling Pt: 138…139 °C (18 mmHg) Density: 1.122 |
MDL Number:
MFCD00009120 CAS Number: 1603-79-8 |
Specification Test Results
Boiling point | 138-139°C/1 |
Density | 1.122 |
ND20 | 1.516 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...