tert-Butyl-1,4-diazepane-1-carboxylate ≥99%

Supplier: Matrix Scientific

Synonyms: 1-BOC-hexahydro-1,4-diazepine, tert-Butyl 1,4-diazepane-1-carboxylate

020023-1G 020023-5G
101809-254EA 36.92 USD
101809-254 101809-256
tert-Butyl-1,4-diazepane-1-carboxylate ≥99%
tert-Butyl-1,4-diazepane-1-carboxylate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 90 °C (0.5 mmHg)
Density: 1.016
MDL Number: MFCD00276987
CAS Number: 112275-50-0

Order Now

Specification Test Results

Boiling point 90°C/0.5mm
Density 1.016
ND20 1.471

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR