Ethyl-4-oxocyclohexanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

012621-25G 012621-100G
101802-994EA 26.15 USD
101802-994 101802-996
Ethyl-4-oxocyclohexanecarboxylate ≥97%
Ethyl-4-oxocyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 150-152°C/40mm
Melting Pt: 221-226°C
Density: 1.068
MDL Number: MFCD00013285
CAS Number: 17159-79-4

Order Now

Specification Test Results

Boiling point 150-152°C/4
Density 1.068
ND20 1.459

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR