Diethyl (bromodifluoromethyl)phosphonate ≥97%
Supplier: Matrix Scientific
007430-25G
007430-100G
101792-748EA
80
USD
101792-748
101792-750
Diethyl (bromodifluoromethyl)phosphonate ≥97%
Diethyl (bromodifluoromethyl)phosphonate
Hazard: Irritant/Irritant-Harmful
Formula:
C₅H₁₀BrF₂O₃P MW: 267.01 g/mol Boiling Pt: 97-98°C/19mm Density: 1.503 |
MDL Number:
MFCD00069118 CAS Number: 65094-22-6 |
Specification Test Results
Boiling point | 97-98°C/19m |
Density | 1.503 |
ND20 | 1.489 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...